missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Periplasmic serine endoprotease DegP Protein
A DNA sequence encoding the Escherichia coli (strain K12) Periplasmic serine endoprotease DegP, was expressed in the hosts and tags indicated.
352.00 € - 3175.00 €
Specifications
Accession Number | NP_414703.1 |
---|---|
Gene ID (Entrez) | 947139 |
Name | Periplasmic serine endoprotease DegP Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16007020
|
enQuireBio™
QP7007-YE-100UG |
100 μg |
1050.00 €
100µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047020
|
enQuireBio™
QP7007-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16017020
|
enQuireBio™
QP7007-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037020
|
enQuireBio™
QP7007-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057020
|
enQuireBio™
QP7007-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027020
|
enQuireBio™
QP7007-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_414703.1 | |
Periplasmic serine endoprotease DegP Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
947139 | |
Research Use Only | |
degP | |
Recombinant Protein | |
Yeast | |
AETSSATTAQ QMPSLAPMLE KVMPSVVSIN VEGSTTVNTP RMPRNFQQFF GDDSPFCQEG SPFQSSPFCQ GGQGGNGGGQ QQKFMALGSG VIIDADKGYV VTNNHVVDNA TVIKVQLSDG RKFDAKMVGK DPRSDIALIQ IQNPKNLTAI KMADSDALRV GDYTVAIGNP FGLGETVTSG IVSALGRSGL NAENYENFIQ TDAAINRGNS GGALVNLNGE LIGINTAILA PDGGNIGIGF AIPSNMVKNL TSQMVEYGQV KRGELGIMGT ELNSELAKAM KVDAQRGAFV SQVLPNSSAA KAGIKAGDVI TSLNGKPISS FAALRAQVGT MPVGSKLTLG LLRDGKQVNV NLELQQSSQN QVDSSSIFNG IEGAEMSNKG KDQGVVVNNV KTGTPAAQIG LKKGDVIIGA NQQAVKNIAE LRKVLDSKPS VLALNIQRGD STIYLLMQ | |
Greater than 90% as determined by SDS-PAGE. |