missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Calpain-14 Protein
A DNA sequence encoding the Homo sapiens (Human) Calpain-14, was expressed in the hosts and tags indicated.
700.00 € - 1955.00 €
Specifications
Accession Number | NP_001138594.1 |
---|---|
Gene ID (Entrez) | 440854 |
Name | Calpain-14 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15932778
|
enQuireBio™
QP10031-BA-20UG |
20 μg |
700.00 €
20µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15922778
|
enQuireBio™
QP10031-BA-100UG |
100 μg |
1955.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942778
|
enQuireBio™
QP10031-BA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001138594.1 | |
Calpain-14 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
440854 | |
Research Use Only | |
CAPN14 | |
Recombinant Protein | |
Baculovirus | |
LFEDTSFPAT LSSIGSGSLL QKLPPRLQWK RPPELHSNPQ FYFAKAKRLD LCQGIVGDCW FLAALQALAL HQDILSRVVP LNQSFTEKYA GIFRFWFWHY GNWVPVVIDD RLPVNEAGQL VFVSSTYKNL FWGALLEKAY AKLSGSYEDL QSGQVSEALV DFTGGVTMTI NLAEAHGNLW DILIEATYNR TLIGCQTHSG EKILENGLVE GHAYTLTGIR KVTCKHRPEY LVKLRNPWGK VEWKGDWSDS SSKWELLSPK EKILLLRKDN DGEFWMTLQD FKTHFVLLVI CKLTPGLLSQ EAAQKWTYTM REGRWEKRST AGGQRQLLQD TFWKNPQFLL SVWRPEEGRR SLRPCSVLVS LLQKPRHRCR KRKPLLAIGF YLYRMNKYHD DQRRLPPEFF QRNTPLSQPD RFLKEKEVSQ ELCLEPGTYL IVPCILEAHQ KSEFVLRVFS RKHIFYEIGS NSGVVFSKEI EDQNERQDEF FTKFFEKHPE INAVQLQNLL NQMTWSSLGS RQPFFSLEAC QGILALLDLN ASGTMSIQEF RDLWKQLKLS QKVFHKQDRG SGYLNWEQLH AAMREAGIML SDDVCQLMLI RYGGPRLQMD FVSFIHLMLR VENMEDVFQN LTQDGKGIYL QKPEWMMMAL YS | |
Greater than 80% as determined by SDS-PAGE. |