missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA repair protein complementing XP-G cells Protein
A DNA sequence encoding the Homo sapiens (Human) DNA repair protein complementing XP-G cells, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_000114.2 |
---|---|
Gene ID (Entrez) | 2073 |
Name | DNA repair protein complementing XP-G cells Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959209
|
enQuireBio™
QP5994-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979209
|
enQuireBio™
QP5994-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15969209
|
enQuireBio™
QP5994-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15989209
|
enQuireBio™
QP5994-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15999209
|
enQuireBio™
QP5994-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15909219
|
enQuireBio™
QP5994-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000114.2 | |
DNA repair protein complementing XP-G cells Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
2073 | |
Research Use Only | |
ERCC5 | |
Recombinant Protein | |
E. coli | |
SFLWGKPDLD KIREFCQRYF GWNRTKTDES LFPVLKQLDA QQTQLRIDSF FRLAQQEKED AKRIKSQRLN RAVTCMLRKE KEAAASEIEA VSVAMEKEFE LLDKAKGKTQ KRGITNTLEE SSSLKRKRLS DSKGKNTCGG FLGETCLSES SDGSSSEDAE SSSLMNVQRR TAAKEPKTSA SDSQNSVKEA PVKNGGATTS SSSDSDDDGG KEKMVLVTAR SVFGKKRRKL RRARGRKRKT | |
Greater than 90% as determined by SDS-PAGE. |