missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF4G1 Protein
A DNA sequence encoding the Homo sapiens (Human) EIF4G1, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_004944.3 |
---|---|
Gene ID (Entrez) | 1981 |
Name | EIF4G1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15989059
|
enQuireBio™
QP5975-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15909069
|
enQuireBio™
QP5975-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15999059
|
enQuireBio™
QP5975-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15919069
|
enQuireBio™
QP5975-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15929069
|
enQuireBio™
QP5975-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15939069
|
enQuireBio™
QP5975-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004944.3 | |
EIF4G1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
1981 | |
Research Use Only | |
EIF4G1 | |
Recombinant Protein | |
E. coli | |
IEEYLHLNDM KEAVQCVQEL ASPSLLFIFV RHGVESTLER SAIAREHMGQ LLHQLLCAGH LSTAQYYQGL YEILELAEDM EIDIPHVWLY LAELVTPILQ EGGVPMGELF REITKPLRPL GKAASLLLEI LGLLCKSMGP KKVGTLWREA GLSWKEFLPE GQDIGAFVAE QKVEYTLGEE SEAPGQRALP SEELNRQLEK LLKEGSSNQR VFDWIEANLS EQQIVSNTLV RALMTAVCYS AIIFETPLRV DVAVLKARAK LLQKYLCDEQ KELQALYALQ ALVVTLEQPP NLLRMFFDAL YDEDVVKEDA FYSWESSKDP AEQQGKGVAL KSVTAFFKWL REAEEESDHN | |
Greater than 90% as determined by SDS-PAGE. |