missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Phenylalanine--tRNA ligase alpha subunit Protein
A DNA sequence encoding the Homo sapiens (Human) Phenylalanine--tRNA ligase alpha subunit, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_004452.1 |
---|---|
Gene ID (Entrez) | 2193 |
Name | Phenylalanine--tRNA ligase alpha subunit Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16013791
|
enQuireBio™
QP8288-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16033791
|
enQuireBio™
QP8288-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023791
|
enQuireBio™
QP8288-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043791
|
enQuireBio™
QP8288-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053791
|
enQuireBio™
QP8288-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16063791
|
enQuireBio™
QP8288-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004452.1 | |
Phenylalanine--tRNA ligase alpha subunit Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
2193 | |
Research Use Only | |
FARSA | |
Recombinant Protein | |
E. coli | |
ADGQVAELLL RRLEASDGGL DSAELAAELG MEHQAVVGAV KSLQALGEVI EAELRSTKHW ELTAEGEEIA REGSHEARVF RSIPPEGLAQ SELMRLPSGK VGFSKAMSNK WIRVDKSAAD GPRVFRVVDS MEDEVQRRLQ LVRGGQAEKL GEKERSELRK RKLLAEVTLK TYWVSKGSAF STSISKQETE LSPEMISSGS WRDRPFKPYN FLAHGVLPDS GHLHPLLKVR SQFRQIFLEM GFTEMPTDNF IESSFWNFDA LFQPQQHPAR DQHDTFFLRD PAEALQLPMD YVQRVKRTHS QGGYGSQGYK YNWKLDEARK NLLRTHTTSA SARALYRLAQ KKPFTPVKYF SIDRVFRNET LDATHLAEFH QIEGVVADHG LTLGHLMGVL REFFTKLGIT QLRFKPAYNP YTEPSMEVFS YHQGLKKWVE VGNSGVFRPE MLLPMGLPEN VSVIAWGLSL ERPTMIKYGI NNIRELVGHK VNLQMVYDSP LCRLDAEPRP PPTQEAA | |
Greater than 90% as determined by SDS-PAGE. |