missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse S100A8 / CAGA Protein
A DNA sequence encoding the Mus musculus (Mouse) S100A8 / CAGA, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_038678.1 |
---|---|
Gene ID (Entrez) | 20201 |
Name | S100A8 / CAGA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024740
|
enQuireBio™
QP6641-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16044740
|
enQuireBio™
QP6641-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16034740
|
enQuireBio™
QP6641-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054740
|
enQuireBio™
QP6641-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064740
|
enQuireBio™
QP6641-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074740
|
enQuireBio™
QP6641-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Spécification
NP_038678.1 | |
S100A8 / CAGA Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
20201 | |
Research Use Only | |
S100A8 | |
Recombinant Protein | |
E. coli | |
PSELEKALSN LIDVYHNYSN IQGNHHALYK NDFKKMVTTE CPQFVQNINI ENLFRELDIN SDNAINFEEF LAMVIKVGVA SHKDSHKE | |
Greater than 90% as determined by SDS-PAGE. |