missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Transmembrane protease serine 4 Protein
A DNA sequence encoding the Mus musculus (Mouse) Transmembrane protease serine 4, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_663378.1 |
---|---|
Gene ID (Entrez) | 214523 |
Name | Transmembrane protease serine 4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16011631
|
enQuireBio™
QP7925-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16031631
|
enQuireBio™
QP7925-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16021631
|
enQuireBio™
QP7925-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041631
|
enQuireBio™
QP7925-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051631
|
enQuireBio™
QP7925-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061631
|
enQuireBio™
QP7925-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_663378.1 | |
Transmembrane protease serine 4 Protein | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
214523 | |
Research Use Only | |
Tmprss4 | |
Recombinant Protein | |
E. coli | |
KVILDKYYFI CGSPLTFIQR GQLCDGHLDC ASGEDEEHCV KDFPEKPGVA VRLSKDRSTL QVLDAATGTW ASVCFDNFTE ALAKTACRQM GYDSQPAFRA VEIRPDQNLP VAQVTGNSQE LQVQNGSRSC LSGSLVSLRC LDCGKSLKTP RVVGGVEAPV DSWPWQVSIQ YNKQHVCGGS ILDPHWILTA AHCFRKYLDV SSWKVRAGSN ILGNSPSLPV AKIFIAEPNP LYPKEKDIAL VKLQMPLTFS GSVRPICLPF SDEVLVPATP VWVIGWGFTE ENGGKMSDML LQASVQVIDS TRCNAEDAYE GEVTAEMLCA GTPQGGKDTC QGDSGGPLMY HSDKWQVVGI VSWGHGCGGP STPGVYTKVT AYLNWIYNVR KSEM | |
Greater than 90% as determined by SDS-PAGE. |