missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig Epididymal secretory glutathione peroxidase Protein
A DNA sequence encoding the Sus scrofa (Pig) Epididymal secretory glutathione peroxidase, was expressed in the hosts and tags indicated.
352.00 € - 3175.00 €
Specifications
Accession Number | NP_999051.1 |
---|---|
Gene ID (Entrez) | 396920 |
Name | Epididymal secretory glutathione peroxidase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16030930
|
enQuireBio™
QP6117-YE-100UG |
100 μg |
1050.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16080930
|
enQuireBio™
QP6117-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16040930
|
enQuireBio™
QP6117-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060930
|
enQuireBio™
QP6117-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070930
|
enQuireBio™
QP6117-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050930
|
enQuireBio™
QP6117-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_999051.1 | |
Epididymal secretory glutathione peroxidase Protein | |
Not determined. | |
Full Length of Mature Protein | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |
396920 | |
Research Use Only | |
GPX5 | |
Recombinant Protein | |
Yeast | |
NSNLEKMDCY KDVTGTIYDY DAFTLNGNEH IQFKQYAGKH VLFVNVATYC GLTAQYPELN TLQEELKPFG LVVLGFPCNQ FGKQEPGENS EILLGLKYVR PGGGYVPNFQ LFEKGDVNGE KEQKVFTFLK HSCPHPSELI GSIGYISWEP IRVHDIRWNF EKFLVGPDGV PVMRWVHETP ISTVKSDILA YLKQFKTE | |
Greater than 90% as determined by SDS-PAGE. |