missing translation for 'onlineSavingsMsg'
Learn More

Reelin Antibody (CL5324) [mFluor Violet 450 SE], R&D Systems™

Product Code. 30297138 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30297138 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30297138 Supplier Novus Biologicals Supplier No. NBP261414MFV450
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Reelin Monoclonal antibody specifically detects Reelin in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Reelin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL5324
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.21, EC 3.4.21.-, reelin, RLPRO1598
Host Species Mouse
Immunogen This antibody was developed using a recombinant protein derived from P78509, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5649
Target Species Human, Mouse, Rat
Content And Storage Store at 4⪚C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.