missing translation for 'onlineSavingsMsg'
Learn More

Reg1A Antibody (431202), FITC, R&D Systems™

Product Code. 18606523 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18606523 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18606523 Supplier Novus Biologicals Supplier No. FAB4937F
  • ONLINE EXCLUSIVE PRICE
    728.45 €
    627.90 € / 0.1mL
    Save 100.55 €
    13% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rat Monoclonal Antibody

Reg1A Monoclonal specifically detects Reg1A in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Reg1A
Applications CyTOF, Flow Cytometry (Intracellular Staining), Western Blot
Classification Monoclonal
Clone 431202
Conjugate FITC
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias ICRF, Islet cells regeneration factor, Islet of Langerhans regenerating protein, lithostathine-1-alpha, P19, pancreatic stone protein, secretory, Pancreatic thread protein, protein-X, PSP, PSPS, PSPS1, PTP, REG, REG-1-alpha, Regenerating islet-derived protein 1-alpha, Regenerating protein I alpha
Host Species Rat
Immunogen E. coli-derived recombinant human Reg1A, Gln23-Asn166, Accession # P05451
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5967
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.