missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Regorafenib Monohydrate, Mikromol™
Description
Regorafenib Monohydrate
Specifications
Specifications
| Chemical Name or Material | Regorafenib Monohydrate |
| CAS | 1019206-88-2 |
| Analyte or Component Names | Regorafenib Monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H15 Cl F4 N4 O3 . H2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Pyridinecarboxamide, 4-[4-[[[[4-chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]-3-fluorophenoxy]-N-methyl-, hydrate (1:1), Regorafenib Monohydrate |
| InChI Formula | InChI=1S/C21H15ClF4N4O3.H2O/c1-27-19(31)18-10-13(6-7-28-18)33-12-3-5-17(16(23)9-12)30-20(32)29-11-2-4-15(22)14(8-11)21(24,25)26;/h2-10H,1H3,(H,27,31)(H2,29,30,32);1H2 |
| SMILES | O.CNC(=O)c1cc(Oc2ccc(NC(=O)Nc3ccc(Cl)c(c3)C(F)(F)F)c(F)c2)ccn1 |
| IUPAC Name | 4-[4-[[4-chloro-3-(trifluoromethyl)phenyl]carbamoylamino]-3-fluorophenoxy]-N-methylpyridine-2-carboxamide;hydrate |
| Mehr anzeigen |
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?