missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Regulatory protein GAL4 Antibody (8C-1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30970-25ug
This item is not returnable.
View return policy
Description
Regulatory protein GAL4 Monoclonal antibody specifically detects Regulatory protein GAL4 in Yeast samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| Regulatory protein GAL4 | |
| Monoclonal | |
| Unconjugated | |
| Phosphate buffered saline | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Yeast | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 8C-1 | |
| Western Blot : 1-10 μg/mL, Immunohistochemistry, Immunoprecipitation | |
| GAL4;GAL81;YPL248C | |
| Hybridoma produced by the fusion of splenocytes from mice immunized with full length Regulatory protein GAL4 expressed in E. Coli and mouse myeloma cells. | |
| 25 μg | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction