missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Regulatory protein GAL4 Antibody (8C-1), Novus Biologicals™
Mouse Monoclonal Antibody
225.00 € - 531.00 €
Specifications
| Antigen | Regulatory protein GAL4 |
|---|---|
| Clone | 8C-1 |
| Dilution | Western Blot : 1-10 μg/mL, Immunohistochemistry, Immunoprecipitation |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067016
|
Novus Biologicals™
NBP3-30970-25ug |
25 μg |
225.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066773
|
Novus Biologicals™
NBP3-30970-100ug |
100 μg |
531.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Regulatory protein GAL4 Monoclonal antibody specifically detects Regulatory protein GAL4 in Yeast samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western BlotSpecifications
| Regulatory protein GAL4 | |
| Western Blot : 1-10 μg/mL, Immunohistochemistry, Immunoprecipitation | |
| Monoclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline | |
| Hybridoma produced by the fusion of splenocytes from mice immunized with full length Regulatory protein GAL4 expressed in E. Coli and mouse myeloma cells. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 8C-1 | |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Yeast | |
| GAL4;GAL81;YPL248C | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title