Learn More
Regulatory protein GAL4 Antibody, Allophycocyanin, R&D Systems™
Product Code. 18603630 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
Product Code. Unit Size
18603630 0.1mL
1 options
Product Code. 18603630 Supplier Novus Biologicals Supplier No. NBP276808APC
  • ONLINE EXCLUSIVE PRICE
    634.33 €
    553.35 € / 0.1mL
    Save 80.98 €
    12% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
Regulatory protein GAL4 Polyclonal specifically detects Regulatory protein GAL4 in Yeast samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Antibody
Product Line Novus