missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Relaxin-3 Antibody (MM0526-7G14) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11866
This item is not returnable.
View return policy
Description
Relaxin-3 Monoclonal specifically detects Relaxin-3 in Human samples. It is validated for Western Blot, Block/Neutralize.
Specifications
| Relaxin-3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000, Block/Neutralize | |
| Q8WXF3 | |
| RLN3 | |
| Human recombinant Relaxin-3 | |
| 0.1 mg | |
| Neuroscience, Neurotransmission | |
| 117579 | |
| Human | |
| Purified |
| Western Blot, Blocking Assay | |
| MM0526-7G14 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
| H3, insl7, relaxin 3, RXN3, ZINS4 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction