missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Relaxin-3 Antibody (MM0526-7G14) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
195.00 € - 510.00 €
Specifications
| Antigen | Relaxin-3 |
|---|---|
| Clone | MM0526-7G14 |
| Concentration | LYOPH |
| Applications | Western Blot, Blocking Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18044397
|
Novus Biologicals
NBP2-11866-0.025MG |
0.025 mg |
195.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18078066
|
Novus Biologicals
NBP2-11866 |
0.1 mg |
510.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Relaxin-3 Monoclonal specifically detects Relaxin-3 in Human samples. It is validated for Western Blot, Block/Neutralize.Specifications
| Relaxin-3 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| H3, insl7, relaxin 3, RXN3, ZINS4 | |
| RLN3 | |
| Protein G purified |
| MM0526-7G14 | |
| Western Blot, Blocking Assay | |
| Unconjugated | |
| Mouse | |
| Neuroscience, Neurotransmission | |
| Q8WXF3 | |
| 117579 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title