missing translation for 'onlineSavingsMsg'
Learn More

Relaxin R1/LGR7 Antibody (933344) [Allophycocyanin], R&D Systems™

Product Code. 18621094 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18621094 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18621094 Supplier Novus Biologicals Supplier No. FAB8898A
  • ONLINE EXCLUSIVE PRICE
    766.31 €
    651.00 € / 0.1mL
    Save 115.31 €
    15% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Relaxin R1/LGR7 Monoclonal specifically detects Relaxin R1/LGR7 in Human, Mouse, Rat samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Relaxin R1/LGR7
Applications CyTOF, Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 933344
Conjugate APC
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation PBS
Gene Alias leucine-rich repeat-containing G protein-coupled receptor 7, Leucine-rich repeat-containing G-protein coupled receptor 7, LGR7.1, LGR7.10, LGR7LGR7.2, MGC138347, MGC142177, Relaxin family peptide receptor 1, relaxin receptor 1, relaxin/insulin-like family peptide receptor 1, RXFPR1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Relaxin R1, Met1-Ser398, Accession # Q9HBX9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 59350
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.