missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-30413SS
This item is not returnable.
View return policy
Description
Respiratory Syncytial Virus Polyclonal specifically detects Respiratory Syncytial Virus in Virus samples. It is validated for Western Blot.
Specifications
| Respiratory Syncytial Virus | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:500 -1:1000 | |
| Fusion glycoprotein F0, Fusion glycoprotein F1, Fusion glycoprotein F2, RS virus, RSV | |
| Whole RSV virions (subgroup A-Long strain) were used as an immunogen. | |
| 0.025 mg | |
| Virology Bacteria and Parasites | |
| Whole RSV virions (subgroup A-Long strain) were used as an immunogen. Detects G, F, N and M2 proteins of RSV. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction