missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
211.00 € - 497.00 €
Specifications
| Antigen | Respiratory Syncytial Virus |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 -1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18243605
|
Novus Biologicals
NBP1-30413SS |
0.025 mg |
211.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18759973
|
Novus Biologicals
NBP1-30413 |
0.1 mg |
497.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Respiratory Syncytial Virus Polyclonal specifically detects Respiratory Syncytial Virus in Virus samples. It is validated for Western Blot.Specifications
| Respiratory Syncytial Virus | |
| Western Blot 1:500 -1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Virus | |
| Fusion glycoprotein F0, Fusion glycoprotein F1, Fusion glycoprotein F2, RS virus, RSV | |
| IgG | |
| Protein G purified | |
| Whole RSV virions (subgroup A-Long strain) were used as an immunogen. Detects G, F, N and M2 proteins of RSV. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Virology Bacteria and Parasites | |
| PBS with 0.02% Sodium Azide | |
| Whole RSV virions (subgroup A-Long strain) were used as an immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title