missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus fusion protein Antibody (10B6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-63020
This item is not returnable.
View return policy
Description
Respiratory Syncytial Virus fusion protein Monoclonal specifically detects Respiratory Syncytial Virus fusion protein in Virus samples. It is validated for Flow Cytometry, ELISA.
Specifications
| Respiratory Syncytial Virus fusion protein | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Recognises the Respiratory Syncytial Virus (RSV) Fusion protein, also known as F-protein. The F-protein is a membrane-associated protein found in the membrane of infected respiratory epithelial cells. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Flow Cytometry, ELISA | |
| 10B6 | |
| Unconjugated | |
| RSV fusion protein | |
| Purified Respiratory Syncytial Virus (RSV) | |
| 0.1 mg | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction