missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus fusion protein Antibody (10B6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
212.00 € - 454.00 €
Specifications
| Antigen | Respiratory Syncytial Virus fusion protein |
|---|---|
| Clone | 10B6 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
| Applications | ELISA, Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18044744
|
Novus Biologicals
NB100-63020-0.025MG |
0.025 mg |
212.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18043306
|
Novus Biologicals
NB100-63020 |
0.1 mg | |||||||
Description
Respiratory Syncytial Virus fusion protein Monoclonal specifically detects Respiratory Syncytial Virus fusion protein in Virus samples. It is validated for Flow Cytometry, ELISA.Specifications
| Respiratory Syncytial Virus fusion protein | |
| 1.0 mg/mL | |
| ELISA, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Virus | |
| Purified Respiratory Syncytial Virus (RSV) | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 10B6 | |
| Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| RSV fusion protein | |
| IgG2b | |
| Protein A or G purified | |
| Recognises the Respiratory Syncytial Virus (RSV) Fusion protein, also known as F-protein. The F-protein is a membrane-associated protein found in the membrane of infected respiratory epithelial cells. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title