missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus M2-1 Protein Antibody, DyLight 405, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Respiratory Syncytial Virus M2-1 Protein Polyclonal antibody specifically detects Respiratory Syncytial Virus M2-1 Protein in Virus samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Respiratory Syncytial Virus M2-1 Protein |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 405 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Fusion glycoprotein F0, Fusion glycoprotein F1, Fusion glycoprotein F2, RS virus, RSV |
| Host Species | Rabbit |
| Immunogen | Reacts with residues 81-95 [LESYIGSINNITKQSA] of the human Respiratory Syncytial Virus M2 protein, which is a CTL epitope. 93% identity to bovine RSV M2. |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?