missing translation for 'onlineSavingsMsg'
Learn More

RFC5 Antibody [DyLight 594], Novus Biologicals Biologicals™

Product Code. 30514759 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30514759 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30514759 Supplier Novus Biologicals Supplier No. NBP338501DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RFC5 Polyclonal antibody specifically detects RFC5 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFC5
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Activator 1 36 kDa subunit, Activator 1 subunit 5,36.5 kD subunit, MGC1155, replication factor C (activator 1) 5, 36.5kDa, RF-C 36 kDa subunit, RFC36replication factor C (activator 1) 5 (36.5kD)
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 181-340 of human RFC5 (NP_031396.1).,, Sequence:, LMVPRLEHVVEEEKVDISEDGMKALVTLSSGDMRRALNILQSTNMAFGKVTEETVYTCTGHPLKSDIANILDWMLNQDFTTAYRNITELKTLKGLALHDILTEIHLFVHRVDFPSSVRIHLLTKMADIEYRLSVGTNEKIQLSSLIAAFQVTRDLIVAEA
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 5985
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.