missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGM-A Antibody (CL11342) [Allophycocyanin], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
RGM-A Monoclonal antibody specifically detects RGM-A in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | RGM-A |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | CL11342 |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | RGM domain family member A, RGM domain family, member A, RGMrepulsive guidance molecule A |
| Host Species | Mouse |
| Immunogen | This antibody was generated using a synthetic peptide of Q96B86, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?