missing translation for 'onlineSavingsMsg'
Learn More

RGM-A Antibody (CL11342) [mFluor Violet 610 SE], Novus Biologicals Biologicals™

Product Code. 30528620 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30528620 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30528620 Supplier Novus Biologicals Supplier No. NBP344194MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RGM-A Monoclonal antibody specifically detects RGM-A in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGM-A
Applications Western Blot
Classification Monoclonal
Clone CL11342
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias RGM domain family member A, RGM domain family, member A, RGMrepulsive guidance molecule A
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of Q96B86, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 56963
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.