missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RGS3 Polyclonal specifically detects RGS3 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
Specifications
| Antigen | RGS3 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunoprecipitation |
| Formulation | PBS (pH 7.2), 1% BSA, 50% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | P49796 |
| Gene Alias | C2PA, FLJ20370, FLJ31516, FLJ90496, PDZ-RGS3, regulator of G-protein signaling 3, regulator of G-protein signalling 3, RGP3 |
| Gene Symbols | RGS3 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?