missing translation for 'onlineSavingsMsg'
Learn More

RGS3 Antibody, Novus Biologicals™

Product Code. p-7199706 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18277662 0.025 mg 25µg
18255100 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18277662 Supplier Novus Biologicals Supplier No. NB12025640.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RGS3 Polyclonal specifically detects RGS3 in Human samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS3
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunoprecipitation
Formulation PBS (pH 7.2), 1% BSA, 50% Glycerol with 0.02% Sodium Azide
Gene Accession No. P49796
Gene Alias C2PA, FLJ20370, FLJ31516, FLJ90496, PDZ-RGS3, regulator of G-protein signaling 3, regulator of G-protein signalling 3, RGP3
Gene Symbols RGS3
Host Species Rabbit
Immunogen E. coli expressed human RGS 3 (amino acids 401-519).
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5998
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.