missing translation for 'onlineSavingsMsg'
Learn More

Rhodopsin Antibody (Rho 1D4) - Chimeric - Azide and BSA Free, R&D Systems™

Product Code. 18612149 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.2mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18612149 0.2 mg 0.2mg
18641419 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18612149 Supplier Novus Biologicals Supplier No. NBP3090010.2mg
  • ONLINE EXCLUSIVE PRICE
    734.59 €
    642.60 € / 0.2mg
    Save 91.99 €
    12% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Rhodopsin Monoclonal specifically detects Rhodopsin in Amphibian∣Bovine∣Human∣Mouse∣Rat∣Zebrafish samples. It is validated for ELISA∣Immunocytochemistry/Immunofluorescence∣Immunohistochemistry∣Immunoprecipitation∣Radioimmune Assays (RIA)∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rhodopsin
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone Rho 1D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS
Gene Alias CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4
Host Species Mouse
Immunogen Bleached bovine rod outer segment (ROS).
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline GPCR, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6010
Target Species Amphibian, Bovine, Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.