missing translation for 'onlineSavingsMsg'
Learn More

Rhot1 Antibody (CL1083) [CoraFluor™ 1], Novus Biologicals™

Product Code. 30297084 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30297084 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30297084

Brand: Novus Biologicals NBP252964CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Rhot1 Monoclonal antibody specifically detects Rhot1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rhot1
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL1083
Conjugate CoraFluor 1
Formulation PBS
Gene Alias ARHT1, EC 3.6.5, EC 3.6.5.-, FLJ11040, FLJ12633, hMiro-1, MIRO-1mitochondrial Rho GTPase 1, mitochondrial Rho 1, rac-GTP binding protein-like protein, Rac-GTP-binding protein-like protein, Ras homolog gene family member T1, ras homolog gene family, member T1
Host Species Mouse
Immunogen This antibody was developed using a recombinant protein derived from Q8IXI2, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55288
Target Species Human
Content And Storage Store at 4⪚C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.