missing translation for 'onlineSavingsMsg'
Learn More

RhoV Antibody, Novus Biologicals™

Product Code. 18677455 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18677455 25 μL 25µL
18684019 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18677455 Supplier Novus Biologicals Supplier No. NBP24908225ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RhoV Polyclonal antibody specifically detects RhoV in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoV
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias ARHV, CDC42-like GTPase 2, CHP, GTP-binding protein-like 2, ras homolog gene family, member V, Rho GTPase-like protein ARHV, rho-related GTP-binding protein RhoV, Wnt-1 regulated Cdc42 homolog 2, Wnt-1 responsive Cdc42 homolog 2, WRCH1-related GTPase, WRCH-2, WRCH2Chp
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: KEVFDSAILSAIEHKARLEKKLNAKGVRTLSRCRWKKFFCFV
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 171177
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.