missing translation for 'onlineSavingsMsg'
Learn More

Ribosome maturation protein SBDS Antibody, Novus Biologicals™

Product Code. 18660354 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18660354 50 μg 50µL
18684012 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18660354 Supplier Novus Biologicals Supplier No. NBP29840450ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Ribosome maturation protein SBDS Polyclonal antibody specifically detects Ribosome maturation protein SBDS in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ribosome maturation protein SBDS
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunoprecipitation 0.1-0.5 μL/mg of lysate, Immunohistochemistry-Paraffin 1:2500-1:15000
Formulation 0.2 um filtered solution in PBS
Gene Alias FLJ10917, ribosome maturation protein SBDS, SDSCGI-97, Shwachman-Bodian-Diamond syndrome, Shwachman-Bodian-Diamond syndrome protein, SWDS
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Mouse Ribosome maturation protein SBDS (Accession#: NP_075737.1; Met1-Glu250)
Purification Method Antigen and protein A Affinity-purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51119
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.