missing translation for 'onlineSavingsMsg'
Learn More

Rift Valley Fever Virus Antibody, R&D Systems™

Product Code. 18604699 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18604699 50 μg 50µL
18672868 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18604699 Supplier Novus Biologicals Supplier No. NBP31275450ul
  • ONLINE EXCLUSIVE PRICE
    342.75 €
    304.50 € / 50µL
    Save 38.25 €
    11% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

Rift Valley Fever Virus Polyclonal specifically detects Rift Valley Fever Virus in Virus samples. It is validated for ELISA∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rift Valley Fever Virus
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias Glycoprotein, RVFV_sM_gp1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Rift Valley Fever Virus (Accession#: AEB20483.1; Met1-Phe250).
Purification Method Antigen and protein A Affinity-purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 9538296
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.