missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Rit1 Polyclonal antibody specifically detects Rit1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Rit1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Phosphate buffered saline |
| Gene Alias | GTP-binding protein Rit1, GTP-binding protein Roc1, MGC125864, MGC125865, Ras-like protein expressed in many tissues, Ras-like without CAAX 1, Ras-like without CAAX protein 1, RIBBRIT, Ric-like, expressed in many tissues, RIT, ROC1Ric (Drosophila)-like, expressed in many tissues |
| Host Species | Rabbit |
| Immunogen | His-tagged recombinant human Rit1 protein |
| Purification Method | Ammonium sulfate precipitation |
| Quantity | 25 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?