missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Rit1 Antibody, Novus Biologicals™

Product Code. p-200086837 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30067061 25 μg 25µg
30067045 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30067061 Supplier Novus Biologicals™ Supplier No. NBP33081125ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Rit1 Polyclonal antibody specifically detects Rit1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rit1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Phosphate buffered saline
Gene Alias GTP-binding protein Rit1, GTP-binding protein Roc1, MGC125864, MGC125865, Ras-like protein expressed in many tissues, Ras-like without CAAX 1, Ras-like without CAAX protein 1, RIBBRIT, Ric-like, expressed in many tissues, RIT, ROC1Ric (Drosophila)-like, expressed in many tissues
Host Species Rabbit
Immunogen His-tagged recombinant human Rit1 protein
Purification Method Ammonium sulfate precipitation
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6016
Target Species Human
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.