missing translation for 'onlineSavingsMsg'
Learn More

RNF34 Antibody [DyLight 650], Novus Biologicals Biologicals™

Product Code. 30516347 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30516347 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30516347 Supplier Novus Biologicals Supplier No. NBP338556C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RNF34 Polyclonal antibody specifically detects RNF34 in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF34
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias CARP1, Caspase regulator CARP1, Caspases-8 and -10-associated RING finger protein 1, E3 ubiquitin-protein ligase RNF34, EC 6.3.2, EC 6.3.2.-, FLJ21786, FYVE-RING finger protein Momo, hRFI, Human RING finger homologous to inhibitor of apoptosis protein, RFI, RIF, RIFF, ring finger protein 34CARP-1, RING finger protein RIFF
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human RNF34 (NP_079402.2).,, Sequence:, MKAGATSMWASCCGLLNEVMGTGAVRGQQSAFAGATGPFRFTPNPEFSTYPPAATEGPNIVCKACGLSFSVFRKKHVCCDCKKDFCSVCSVLQENLRRCSTCHLLQETAFQRPQLMRLKVKDLRQYLILRNIPIDTCREKEDLVDLVLCHHGLGSEDDMDTSSLNSSRSQTSSFFTRSFFSNYTAPSATMSSFQGELMDGDQTSRSGVPAQVQSEITSAN
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 80196
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.