missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF40 Recombinant Rabbit Monoclonal Antibody (24GB1100)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557698
This item is not returnable.
View return policy
Description
RNF40 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The protein encoded by this gene contains a RING finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein was reported to interact with the tumor suppressor protein RB1. Studies of the rat counterpart suggested that this protein may function as an E3 ubiquitin-protein ligase, and facilitate the ubiquitination and degradation of syntaxin 1, which is an essential component of the neurotransmitter release machinery.
Specifications
| RNF40 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| RNF40 | |
| 95 kDa retinoblastoma protein binding protein; 95 kDa retinoblastoma-associated protein; AI848422; BRE1 E3 ubiquitin ligase homolog B; Bre1b; BRE1-B; DKFZp686K191; E3 ubiquitin-protein ligase BRE1B; KIAA0661; MGC13051; mKIAA0661; protein staring; Rb-associated protein; RBP95; RING finger protein 40; ring finger protein 40, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase BRE1B; Rnf40; STARING; staring protein; syntaxin-1-interacting RING finger protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 233900, 266712, 9810 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 24GB1100 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75150, Q3U319, Q8CJB9 | |
| RNF40 | |
| A synthesized peptide derived from human RNF40 (aa 220-450). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction