missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNF40 Recombinant Rabbit Monoclonal Antibody (24GB1100)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

198.00 € - 564.00 €

Specifications

Antigen RNF40
Clone 24GB1100
Content And Storage -20°C
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275189
GREENER_CHOICE
View Documents
Invitrogen™
MA557698
100 μL
564.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30276605
View Documents
Invitrogen™
MA557699
20 μL
198.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RNF40 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

The protein encoded by this gene contains a RING finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein was reported to interact with the tumor suppressor protein RB1. Studies of the rat counterpart suggested that this protein may function as an E3 ubiquitin-protein ligase, and facilitate the ubiquitination and degradation of syntaxin 1, which is an essential component of the neurotransmitter release machinery.
Specifications

Specifications

RNF40
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
RNF40
95 kDa retinoblastoma protein binding protein; 95 kDa retinoblastoma-associated protein; AI848422; BRE1 E3 ubiquitin ligase homolog B; Bre1b; BRE1-B; DKFZp686K191; E3 ubiquitin-protein ligase BRE1B; KIAA0661; MGC13051; mKIAA0661; protein staring; Rb-associated protein; RBP95; RING finger protein 40; ring finger protein 40, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase BRE1B; Rnf40; STARING; staring protein; syntaxin-1-interacting RING finger protein
RNF40
Primary
Affinity Chromatography
24GB1100
Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O75150, Q3U319, Q8CJB9
233900, 266712, 9810
A synthesized peptide derived from human RNF40 (aa 220-450).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.