missing translation for 'onlineSavingsMsg'
Learn More

Robenidine hydrochloride 100 μg/mL in Acetone:Methanol, Dr. Ehrenstorfer

Product Code. 30321497
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321497 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321497 Supplier Dr. Ehrenstorfer Supplier No. DREA16815400AM100
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Robenidine hydrochloride
CAS 25875-50-7
Analyte or Component Names Robenidine hydrochloride
Shipping Condition Room Temperature
Molecular Formula C15 H13 Cl2 N5 . Cl H
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 2,2'-Bis[(4-chlorophenyl)methylene]carbonimidic Dihydrazide Hydrochloride (1:1), Carbonimidic dihydrazide, bis[(4-chlorophenyl)methylene]-, monohydrochloride (9CI), Guanidine, 1,3-bis[(p-chlorobenzylidene)amino]-, monohydrochloride (8CI), 1,3-Bis[(p-chlorobenzylidene)amino]guanidine monohydrochloride, Cycostat, NCL 812, Robenidine hydrochloride, Robenzidene
InChI Formula InChI=1S/C15H13Cl2N5.ClH/c16-13-5-1-11(2-6-13)9-19-21-15(18)22-20-10-12-3-7-14(17)8-4-12;/h1-10H,(H3,18,21,22);1H/b19-9+,20-10+;
SMILES Cl.Clc1ccc(\C=N\NC(=N)N\N=C\c2ccc(Cl)cc2)cc1
IUPAC Name 1,3-bis[(E)-(4-chlorophenyl)methylideneamino]guanidine;hydrochloride
Solvent or Matrix Acetone/Methanol
Molecular Weight (g/mol) 370.66
Quantity 1 mL
Formula Weight 369.0315 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.