missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ropinirole Hydrochloride, Mikromol™
Description
Ropinirole Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Ropinirole Hydrochloride |
| CAS | 91374-20-8 |
| Analyte or Component Names | Ropinirole Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H24 N2 O . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | 2H-Indol-2-one, 4-[2-(dipropylamino)ethyl]-1,3-dihydro-, hydrochloride (1:1), 2H-Indol-2-one, 4-[2-(dipropylamino)ethyl]-1,3-dihydro-, monohydrochloride (9CI), Adartrel, Parkirop, Ralnea XL, Repreve, Requip, Ronirol, Ropark, Ropinirole hydrochloride, Ropitar, SKF 101468 hydrochloride, SKF 101468A, Spiroco XL, SKF-101468-A, Ropinirole hydrochloride (SKF-101468-A), SB-270341, RRT = 1.4 [SB-410189], SKF-98959, RRT = 28.5 [SB-381331], SKF-96413, SB 221243A (4-[2-(Dipropylamino)ethyl]-1-hydro-3-methenyl-2H-indol-2-on monohydrochloride) |
| InChI Formula | InChI=1S/C16H24N2O.ClH/c1-3-9-18(10-4-2)11-8-13-6-5-7-15-14(13)12-16(19)17-15;/h5-7H,3-4,8-12H2,1-2H3,(H,17,19);1H |
| SMILES | Cl.CCCN(CCC)CCc1cccc2NC(=O)Cc12 |
| IUPAC Name | 4-[2-(dipropylamino)ethyl]-1,3-dihydroindol-2-one;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?