missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97604-100ul
This item is not returnable.
View return policy
Description
ROR beta Polyclonal antibody specifically detects ROR beta in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ROR beta | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| NR1F2nuclear receptor ROR-beta, Nuclear receptor RZR-beta, Nuclear receptor subfamily 1 group F member 2, RAR-related orphan receptor B, RAR-related orphan receptor beta, retinoic acid-binding receptor beta, retinoid-related orphan receptor beta, Retinoid-related orphan receptor-beta, ROR-BETA, RZRBbA133M9.1, RZR-BETA | |
| Produced in rabbits immunized with E. coli-derived Human ROR beta fragment. | |
| 100 μg | |
| Cancer, Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction | |
| 6096 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction