missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
227.00 € - 335.00 €
Specifications
| Antigen | ROR beta |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18680722
|
Novus Biologicals
NBP2-97604-100ul |
100 μg |
335.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640164
|
Novus Biologicals
NBP2-97604-50ul |
50 μg |
227.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ROR beta Polyclonal antibody specifically detects ROR beta in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| ROR beta | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction | |
| PBS (pH 7.0) | |
| 6096 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| NR1F2nuclear receptor ROR-beta, Nuclear receptor RZR-beta, Nuclear receptor subfamily 1 group F member 2, RAR-related orphan receptor B, RAR-related orphan receptor beta, retinoic acid-binding receptor beta, retinoid-related orphan receptor beta, Retinoid-related orphan receptor-beta, ROR-BETA, RZRBbA133M9.1, RZR-BETA | |
| Produced in rabbits immunized with E. coli-derived Human ROR beta fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title