missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPA70 Recombinant Rabbit Monoclonal Antibody (23GB4885), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553828
This item is not returnable.
View return policy
Description
This gene Plays an essential role in several cellular processes in DNA metabolism including replication, recombination and DNA repair. Binds and subsequently stabilizes single-stranded DNA intermediates and thus prevents complementary DNA from reannealing.
Specifications
| RPA70 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| RPA1 | |
| 5031405K23Rik; 70kDa; AA589576; AW557552; Cb1-727; HSSB; MST075; MSTP075; REPA1; Replication factor A protein 1; replication protein A 70 kDa DNA-binding subunit; Replication protein A 70 kDa DNA-binding subunit, N-terminally processed; replication protein A1; replication protein A1, 70kDa; RF-A; RF-A protein 1; Rpa; RP-A; RP-A p70; RPA1; RPA70; Single-stranded DNA-binding protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 287524, 6117 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4885 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P27694 | |
| RPA1 | |
| A synthesized peptide derived from human RPA70 (1-54AA). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction