missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPA70 Recombinant Rabbit Monoclonal Antibody (23GB4885), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
207.00 € - 562.00 €
Specifications
| Antigen | RPA70 |
|---|---|
| Clone | 23GB4885 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089624
![]() |
Invitrogen™
MA553828 |
100 μL |
562.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089597
|
Invitrogen™
MA553829 |
20 μL |
207.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPA70 Recombinant Monoclonal antibody specifically detects RPA70 in Human, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| RPA70 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| P27694 | |
| 287524, 6117 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4885 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 5031405K23Rik; 70kDa; AA589576; AW557552; Cb1-727; HSSB; MST075; MSTP075; REPA1; Replication factor A protein 1; replication protein A 70 kDa DNA-binding subunit; Replication protein A 70 kDa DNA-binding subunit, N-terminally processed; replication protein A1; replication protein A1, 70kDa; RF-A; RF-A protein 1; Rpa; RP-A; RP-A p70; RPA1; RPA70; Single-stranded DNA-binding protein | |
| A synthesized peptide derived from human RPA70 (1-54AA). | |
| Primary | |
| -20°C | |
| RPA1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title