missing translation for 'onlineSavingsMsg'
Learn More

(RS)-Equol, LoGiCal

Product Code. 30461863
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461863 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461863 Supplier LoGiCal Supplier No. LGCFOR0747.02
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

(RS)-Equol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material R,S-Equol
CAS 94105-90-5
Analyte or Component Names R,S-Equol
Shipping Condition Room Temperature
Molecular Formula C15 H14 O3
Certifications/Compliance ISO 9001
Synonym 2 H-1-Benzopyran-7-ol, 3,4-dihydro-3-(4-hydroxyphenyl)-, 7,4'-Homoisoflavane, (+/-)-Equol, NV 07 alpha
InChI Formula InChI=1 S/C15H14O3/c16-13-4-1-10(2-5-13)12-7-11-3-6-14(17)8-15(11)18-9-12/h1-6,8,12,16-17 H,7,9H2
SMILES Oc1ccc(cc1)C2COc3cc(O)ccc3C2
IUPAC Name 3-(4-hydroxyphenyl)-3,4-dihydro-2 H-chromen-7-ol
Industry Type Forensics
Molecular Weight (g/mol) 242.27
Quantity 10 mg
Formula Weight 242.0943 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.