missing translation for 'onlineSavingsMsg'
Learn More

Rupatadine (5-Methyl-3-pyridinyl)methyl)pyridinium Chloride Trihydrochloride, TRC

Product Code. 30477521
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477521 10 mg 10mg
30472184 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477521 Supplier Toronto Research Chemicals Supplier No. TRCM32674210MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Rupatadine ((5-Methyl-3-pyridinyl)methyl)pyridinium Chloride Trihydrochloride
Analyte or Component Names Rupatadine ((5-Methyl-3-pyridinyl)methyl)pyridinium Chloride Trihydrochloride
Shipping Condition Room Temperature
Molecular Formula C33H34Cl2N4 . 3(HCl)
Solution Type Neat
SMILES ClC1=CC(CCC2=C/3N=CC=C2)=C(C=C1)C3=C4CCN(CC5=C[N+](CC6=CN=CC(C)=C6)=CC(C)=C5)CC\4.[3 HCl].[Cl-]
Industry Type Pharmaceutical
Molecular Weight (g/mol) 666.94
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.