missing translation for 'onlineSavingsMsg'
Learn More

Rutin trihydrate 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30320846
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320846 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320846 Supplier Dr. Ehrenstorfer Supplier No. DREA16880000AL1000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Rutin hydrate
CAS 250249-75-3
Analyte or Component Names Rutin hydrate
Shipping Condition +20°C
Molecular Formula C27 H30 O16 . 3 H2 O
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Rutoside trihydrate, 3-[[6-O-(6-Deoxy-α-l-mannopyranosyl)-β-d-glucopyranosyl]oxy]-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-4H-1-benzopyran-4-one, 4H-1-Benzopyran-4-one, 3-[[6-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-, hydrate (1:3), 4H-1-Benzopyran-4-one, 3-[[6-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-, trihydrate (9CI), Rutin trihydrate
InChI Formula InChI=1S/C27H30O16.3H2O/c1-8-17(32)20(35)22(37)26(40-8)39-7-15-18(33)21(36)23(38)27(42-15)43-25-19(34)16-13(31)5-10(28)6-14(16)41-24(25)9-2-3-11(29)12(30)4-9;;;/h2-6,8,15,17-18,20-23,26-33,35-38H,7H2,1H3;3*1H2/t8-,15+,17-,18+,20+,21-,22+,23+,26+,27-;;;/m0.../s1
SMILES O.O.O.C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(Oc4cc(O)cc(O)c4C3=O)c5ccc(O)c(O)c5)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O
IUPAC Name 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one;trihydrate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 664.56
Quantity 1 mL
Formula Weight 664.1851 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.