missing translation for 'onlineSavingsMsg'
Learn More

RXR alpha/NR2B1 Antibody (S04-6C3), Novus Biologicals™

Product Code. 18633155 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18633155 100 μg 100µL
18691555 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18633155 Supplier Novus Biologicals Supplier No. NBP319972100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

RXR alpha/NR2B1 Monoclonal antibody specifically detects RXR alpha/NR2B1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RXR alpha/NR2B1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone S04-6C3
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias FLJ00280, FLJ00318, FLJ16020, MGC102720, NR2B1FLJ16733, Nuclear receptor subfamily 2 group B member 1, retinoic acid receptor RXR-alpha, Retinoid X receptor alpha, retinoid X receptor, alpha
Host Species Rabbit
Immunogen A synthetic peptide of human RXR alpha/NR2B1 (Uniprot # P19793)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Lipid and Metabolism, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6256
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.