missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 97%

Product Code. 11446716
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446716 1 g 1g
11456716 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446716 Supplier Thermo Scientific Alfa Aesar Supplier No. B23872.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 76189-56-5 | C44H32P2 | 622.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76189-56-5
Molecular Formula C44H32P2
Molecular Weight (g/mol) 622.69
MDL Number MFCD00010805
InChI Key MUALRAIOVNYAIW-UHFFFAOYSA-N
Synonym binap, r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl, 2,2'-bis diphenylphosphino-1,1'-binaphthyl, s-binap, s---binap, s---2,2'-bis diphenylphosphino-1,1'-binaphthyl, r-+-binap, r-binap, rac-binap, 2,2'-bis diphenylphosphino-1,1'-binaphthalene
PubChem CID 634876
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 239°C to 241°C
Quantity 1 g
Beilstein 5321444
Sensitivity Air Sensitive
Merck Index 14,1223
Optical Rotation −230° (c=0.3 in Toluene)
IUPAC Name [1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane
Formula Weight 622.69
Percent Purity 97%
Chemical Name or Material (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.