missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Hydroxy-3-methoxy-3,3-diphenylpropionic Acid, Mikromol™

Product Code. 30051446
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051446 100 mg 100mg
30050612 25 mg 25mg
30461161 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30051446 Supplier Mikromol Supplier No. MM3756.01
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(S)-2-Hydroxy-3-methoxy-3,3-diphenylpropionic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-2-Hydroxy-3-methoxy-3,3-diphenylpropionic Acid
CAS 178306-52-0
Analyte or Component Names (S)-2-Hydroxy-3-methoxy-3,3-diphenylpropionic Acid
Shipping Condition Room Temperature
Molecular Formula C16 H16 O4
Certifications/Compliance ISO 17025
Synonym Benzenepropanoic acid, α-hydroxy-β-methoxy-β-phenyl-, (S)-, (S)-2-Hydroxy-3-methoxy-3,3-diphenylpropionic acid, 2S-2-Hydroxy-3-methoxy-3,3-diphenylpropanoic acid
InChI Formula InChI=1S/C16H16O4/c1-20-16(14(17)15(18)19,12-8-4-2-5-9-12)13-10-6-3-7-11-13/h2-11,14,17H,1H3,(H,18,19)/t14-/m1/s1
SMILES COC([C@H](O)C(=O)O)(c1ccccc1)c2ccccc2
IUPAC Name (2S)-2-hydroxy-3-methoxy-3,3-diphenylpropanoic acid
Molecular Weight (g/mol) 272.3
Quantity 100 mg
Formula Weight 272.1049 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.