Learn More
S100A1 Antibody (049), Alexa Fluor™ 532 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90504AF532
Product Code. 18415429
  • 660.00 € / 0.10mL
Description
S100A1 Monoclonal antibody specifically detects S100A1 in Mouse samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
S100A1
Monoclonal
Alexa Fluor 532
protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse S100A1 (P56565; Gly2-Ser94).
0.1 mL
Breast Cancer, Cancer, Cell Biology, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Peroxisome Markers, Signal Transduction
6271
Store at 4°C in the dark.
IgG
ELISA
049
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Mouse
Purified